missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSME4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18799-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is expected to recognize all 4 isoforms of PSME4 but appears to mainly detect isoform 2 of 140 kDa.
proteasome (prosome, macropain) activator subunit 4 (PSME4) is subunit of the proteasome. PSME4 has been found to activate proteasomal cleavage of peptides in an energy-independent manner and may be involved in spermatogenesis. PSME4 may also be involved in DNA repair.Specifications
| PSME4 | |
| Polyclonal | |
| Unconjugated | |
| PSME4 | |
| KIAA0077, PA200, Proteasome activator PA200, PSME4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 103554, 23198, 498433 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14997, Q5SSW2 | |
| Psme4 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction