missing translation for 'onlineSavingsMsg'
Learn More

PSME4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen PSME4
Concentration 0.13 mg/mL
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16898634
View Documents
Proteintech
18799-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16888634
View Documents
Proteintech
18799-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The antibody is expected to recognize all 4 isoforms of PSME4 but appears to mainly detect isoform 2 of 140 kDa.

proteasome (prosome, macropain) activator subunit 4 (PSME4) is subunit of the proteasome. PSME4 has been found to activate proteasomal cleavage of peptides in an energy-independent manner and may be involved in spermatogenesis. PSME4 may also be involved in DNA repair.
Specifications

Specifications

PSME4
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
Q14997, Q5SSW2
103554, 23198, 498433
Peptide
Primary
-20°C
PSME4
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
KIAA0077, PA200, Proteasome activator PA200, PSME4
Psme4
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.