missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMB5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19178-1-AP-20UL
This item is not returnable.
View return policy
Description
Cyclic nucleotide phosphodiesterases.Specifications
| PSMB5 | |
| Polyclonal | |
| Unconjugated | |
| PSMB5 | |
| 20S proteasome, LMPX, Macropain epsilon chain, MB1, Proteasome chain 6, Proteasome epsilon chain, Proteasome subunit beta type 5, Proteasome subunit MB1, Proteasome subunit X, PSMB5, X | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19173, 29425, 5693 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O55234, P28074, P28075 | |
| Psmb5 | |
| PSMB5 Fusion Protein Ag5828 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction