Learn More
PSMA4 Mouse, anti-Human, CoraLite Plus 488, Clone: 2E6E4, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | PSMA4 |
|---|---|
| Clone | 2E6E4 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18056415
|
Proteintech
CL488-68203-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18087615
|
Proteintech
CL488-68203-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PSMA4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PSMA4. This antibody recognizes human, pig, rabbit, rat, mouse antigen. The PSMA4 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| PSMA4 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 5685 | |
| IgG2b | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 2E6E4 | |
| Immunofluorescence | |
| Liquid | |
| Human, Pig, Rabbit, Rat, Mouse | |
| HC9, HsT17706, Macropain subunit C9, Proteasome component C9, Proteasome subunit L, PSC9, PSMA4 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.