Learn More
PSMA4 Mouse, anti-Human, CoraLite Plus 488, Clone: 2E6E4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68203-100UL
Description
The PSMA4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PSMA4. This antibody recognizes human, pig, rabbit, rat, mouse antigen. The PSMA4 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| PSMA4 | |
| 2E6E4 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Pig, Rabbit, Rat, Mouse | |
| 155 kDa | |
| IgG2b |
| Immunofluorescence | |
| 1000 μg/mL | |
| HC9, HsT17706, Macropain subunit C9, Proteasome component C9, Proteasome subunit L, PSC9, PSMA4 | |
| 100 μL | |
| 5685 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |