missing translation for 'onlineSavingsMsg'
Learn More

PSMA4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£190.00 - £255.00

Specifications

Antigen PSMA4
Dilution Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:200-1:1000
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18623496
View Documents
Novus Biologicals
NBP2-99610-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18691266
View Documents
Novus Biologicals
NBP2-99610-50ul
50 μg
£190.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PSMA4 Polyclonal antibody specifically detects PSMA4 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications

Specifications

PSMA4
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Proteases & Other Enzymes
PBS (pH 7.0)
5685
IgG
Antigen and protein A Affinity-purified
Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:200-1:1000
Polyclonal
Purified
RUO
Human
HC9EC 3.4.25.1, HsT17706, Macropain subunit C9, MGC12467, MGC24813, Multicatalytic endopeptidase complex subunit C9, proteasome (prosome, macropain) subunit, alpha type, 4, Proteasome component C9, proteasome subunit alpha type-4, proteasome subunit HC9, Proteasome subunit L, PSC9MGC111191
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PSMA4.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.