missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99610-100ul
This item is not returnable.
View return policy
Description
PSMA4 Polyclonal antibody specifically detects PSMA4 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| PSMA4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| HC9EC 3.4.25.1, HsT17706, Macropain subunit C9, MGC12467, MGC24813, Multicatalytic endopeptidase complex subunit C9, proteasome (prosome, macropain) subunit, alpha type, 4, Proteasome component C9, proteasome subunit alpha type-4, proteasome subunit HC9, Proteasome subunit L, PSC9MGC111191 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PSMA4. | |
| 100 μg | |
| Proteases & Other Enzymes | |
| 5685 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction