missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSF™ Black Phylon JCB Low Cut Hikers
Brand: PSF Progressive Safety TREKKER-6
Code : NEW
Additional Details : Weight : 2.00000kg
Specifications
EN ISO 20345:2011, S1 and SRC Standard | |
Phylon | |
6 | |
Rubber, Steel | |
Water and shocks | |
JCB Low Cut Hiker |
Black | |
Phylon | |
Made up of Rubber, Steel midsole | |
Hiker | |
Yes |