missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pseudomonas C-N Selective Agar (Cetrimide Agar)

Product Code. 11914122
Change view
Click to view available options
Fill Volume:
17 mL
19 mL
Fill Weight:
17g ± 5%
19g ± 5%
Unit Size:
Each
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Volume Fill Weight Unit Size
11914122 17 mL 17g ± 5% Pack of 10
12759060 19 mL 19g ± 5% Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11914122 Supplier Thermo Scientific™ Supplier No. PO5076A
  • ONLINE EXCLUSIVE PRICE
    £11.40
    £10.81 / Pack of 10
    Save £0.59
    5% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selectively isolate and differentiate Pseudomonas aeruginosa using Thermo Scientific™ Pseudomonas C-N Selective Agar (Cetrimide Agar) prepared plates.

Selectively isolate and differentiate Pseudomonas aeruginosa using Thermo Scientific™ Pseudomonas C-N Selective Agar (Cetrimide Agar) prepared plates. Pseudomonas aeruginosa produces a characteristic bright green color on the medium. The medium has been tested in accordance with the methods described in ISO 7704:2023.

Pseudomonas C-N Selective Agar (Cetrimide Agar) is for the selective isolation of Pseudomonas aeruginosa from a range of samples.

The formula was described by Goto and Enomoto1 following the demonstration of cetrimide as a selective agent by Lowbury and Collins2.

Cetrimide is a quarternary ammonium compound with bactericidal activity against a broad range of Gram-positive organisms and some Gram-negative organisms.

Goto and Enomoto showed that addition of nalidixic acid at 15μg/mL, while at the same time reducing the cetrimide content to 200mg, improved performance. The medium gave better recovery of Pseudomonas aeruginosa with enhanced pigment formation while strongly suppressing Klebsiella, Proteus and Providencia spp., the latter organisms being the troublesome contaminants of conventional cetrimide media.

Pseudomonas aeruginosa produces a number of water soluble iron chelators, including the yellow-green or yellow-brown fluorescent pyoverdin. When pyoverdin combines with the blue water-soluble pyocyanin, also produced by the organism, the bright green color characteristic of Pseudomonas aeruginosa is produced. The addition of magnesium chloride and potassium sulphate in the medium enhances the production of these chelators.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Color Oyster white, transparent
Content And Storage 2° to 12°C, in the dark
Description Pseudomonas C-N Selective Agar (Cetrimide Agar)
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Volume 17 mL
Fill Weight 17g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.

For professional use only (in vitro diagnostic use).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.