missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSD93 Antibody (S18-30), Novus Biologicals™
Mouse Monoclonal Antibody
£171.00
Specifications
| Antigen | PSD93 |
|---|---|
| Clone | S18-30 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:1000, Microarray |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18792994
|
Novus Biologicals
NBP2-12912-0.025mg |
0.025 mg | |||||||
Description
PSD93 Monoclonal specifically detects PSD93 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| PSD93 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| 1740 | |
| Fusion protein amino acids 1-852 of rat Chapsyn-110 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S18-30 | |
| Western Blot 1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:1000, Microarray | |
| Monoclonal | |
| Purified | |
| RUO | |
| Chapsyn-110, discs, large homolog 2 (Drosophila), discs, large homolog 2, chapsyn-110, discs, large homolog 2, chapsyn-110 (Drosophila), disks large homolog 2, DKFZp781D1854, DKFZp781E0954, FLJ37266, MGC131811, Postsynaptic density protein PSD-93, PSD-93, PSD93,110kDa | |
| DLG2 | |
| IgG1 | |
| Protein G purified | |
| Detects approx 110kDa. No cross-reactivity against other MAGUK family members, PSD95, SAP97, SAP102, expressed in transfected cells. Weak human detection. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title