missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSD93 Antibody (S18-30), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12912-0.025mg
This item is not returnable.
View return policy
Description
PSD93 Monoclonal specifically detects PSD93 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| PSD93 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:1000, Microarray | |
| DLG2 | |
| Fusion protein amino acids 1-852 of rat Chapsyn-110 | |
| 0.025 mg | |
| Primary | |
| Detects approx 110kDa. No cross-reactivity against other MAGUK family members, PSD95, SAP97, SAP102, expressed in transfected cells. Weak human detection. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| S18-30 | |
| Unconjugated | |
| Chapsyn-110, discs, large homolog 2 (Drosophila), discs, large homolog 2, chapsyn-110, discs, large homolog 2, chapsyn-110 (Drosophila), disks large homolog 2, DKFZp781D1854, DKFZp781E0954, FLJ37266, MGC131811, Postsynaptic density protein PSD-93, PSD-93, PSD93,110kDa | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 1740 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction