missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSD-95 Antibody (6G6-1C9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£174.00
Specifications
| Antigen | PSD-95 |
|---|---|
| Clone | 6G6-1C9 |
| Concentration | 3.3 mg/mL |
| Dilution | Western Blot 1:2000, Flow Cytometry 2 μg/ test (100ul), Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Block/Neutralize |
| Applications | Western Blot |
Description
PSD-95 Monoclonal specifically detects PSD-95 in Human, Mouse, Rat, Invertebrate, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Block/Neutralize.Specifications
| PSD-95 | |
| 3.3 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| discs, large homolog 4 (Drosophila), disks large homolog 4, FLJ97752, FLJ98574, post-synaptic density protein 95, Postsynaptic density protein 95, PSD-95, PSD95SAP90SAP-90discs large homolog 4, Synapse-associated protein 90, Tax interaction protein 15 | |
| DLG4 | |
| IgG2a | |
| Protein A purified | |
| Detects Post Synaptic Density 95 kDa (PSD-95) from rat tissues. |
| 6G6-1C9 | |
| Western Blot 1:2000, Flow Cytometry 2 μg/ test (100ul), Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Block/Neutralize | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 1742 | |
| Purified recombinant rat PSD-95. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title