missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSD-95 Antibody (6G6-1C9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-556-0.025ml
This item is not returnable.
View return policy
Description
PSD-95 Monoclonal specifically detects PSD-95 in Human, Mouse, Rat, Invertebrate, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Block/Neutralize.
Specifications
| PSD-95 | |
| Monoclonal | |
| 3.3 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 2 μg/ test (100ul), Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Block/Neutralize | |
| DLG4 | |
| Purified recombinant rat PSD-95. | |
| 0.025 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 1742 | |
| Human | |
| Purified |
| Western Blot | |
| 6G6-1C9 | |
| Unconjugated | |
| discs, large homolog 4 (Drosophila), disks large homolog 4, FLJ97752, FLJ98574, post-synaptic density protein 95, Postsynaptic density protein 95, PSD-95, PSD95SAP90SAP-90discs large homolog 4, Synapse-associated protein 90, Tax interaction protein 15 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Detects Post Synaptic Density 95 kDa (PSD-95) from rat tissues. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction