missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PSC Definitive Endoderm Induction Kit

Product Code. 17164699
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17164699 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17164699 Supplier Gibco™ Supplier No. A3062601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gibco™ PSC Definitive Endoderm Induction Kit consists of two xeno-free media that enable efficient induction of human pluripotent stem cells (PSCs) to definitive endoderm.

Gibco™ PSC Definitive Endoderm Induction Kit consists of two xeno-free media that enable efficient induction of human pluripotent stem cells (PSCs) to definitive endoderm. Unlike other methods that require multiple components and take five or more days, the PSC Definitive Endoderm Induction Kit can generate ≥90% CXCR4+/PDGFRα- definitive endoderm cells with only two components in just two days. Each medium is supplied as a 1X complete medium, requiring no mixing in of additional components, and the resultant definitive endoderm is capable of differentiating into midgut/hindgut, pancreatic endoderm, or liver bud progenitors that express relevant physiological markers.

Generation of high quality definitive endodermDefinitive endoderm generated using the PSC Definitive Endoderm Induction Kit is capable of differentiating into downstream lineages such as midgut/hindgut (expressing CDX2), pancreatic endoderm (expressing PDX2), and liver bud progenitors (expressing AFP).Rapid production of definitive endodermThe PSC Definitive Endoderm Induction Kit is capable of producing definitive endoderm in only two days.Generation of highly pure definitive endodermThe PSC Definitive Endoderm Induction Kit is capable of producing highly pure populations of definitive endoderm from pluripotent stem cells, showing ≥90% CXCR4+/PDGFRα- across multiple lines and high expression of key markers SOX17 and FOXA2.No mixing in of components requiredEach medium is supplied at a 1X concentration, requiring no mixing in of reagents. Simply thaw the media and add to PSC culture.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Stem Cells (Human)
Content And Storage • PSC Definitive Endoderm Induction Medium A
• PSC Definitive Endoderm Induction Medium B
Product Type PSC Definitive Endoderm Induction Kit
Quantity 1 kit
Shipping Condition Dry Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.