missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRRG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | PRRG1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
PRRG1 Polyclonal specifically detects PRRG1 in Human samples. It is validated for Western Blot.Specifications
| PRRG1 | |
| Polyclonal | |
| Rabbit | |
| O14668 | |
| 5638 | |
| Synthetic peptides corresponding to PRRG1(proline rich Gla (G-carboxyglutamic acid) 1) The peptide sequence was selected from the N terminal of PRRG1. Peptide sequence MGRVFLTGEKANSILKRYPRANGFFEEIRQGNIERECKEEFCTFEEAREA. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PRGP1TMG1, proline rich Gla (G-carboxyglutamic acid) 1, Proline-rich gamma-carboxyglutamic acid protein 1, proline-rich Gla (G-carboxyglutamic acid) polypeptide 1, Proline-rich Gla protein 1, transmembrane gamma-carboxyglutamic acid protein 1 | |
| PRRG1 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title