missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRRG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59906
This item is not returnable.
View return policy
Description
PRRG1 Polyclonal specifically detects PRRG1 in Human samples. It is validated for Western Blot.
Specifications
| PRRG1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| PRGP1TMG1, proline rich Gla (G-carboxyglutamic acid) 1, Proline-rich gamma-carboxyglutamic acid protein 1, proline-rich Gla (G-carboxyglutamic acid) polypeptide 1, Proline-rich Gla protein 1, transmembrane gamma-carboxyglutamic acid protein 1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5638 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O14668 | |
| PRRG1 | |
| Synthetic peptides corresponding to PRRG1(proline rich Gla (G-carboxyglutamic acid) 1) The peptide sequence was selected from the N terminal of PRRG1. Peptide sequence MGRVFLTGEKANSILKRYPRANGFFEEIRQGNIERECKEEFCTFEEAREA. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Canine: 92%; Mouse: 92%; Rabbit: 92%; Rat: 92%; Xenopus: 91%; Guinea pig: 85%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction