missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPF6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57544
This item is not returnable.
View return policy
Description
PRPF6 Polyclonal specifically detects PRPF6 in Human samples. It is validated for Western Blot.
Specifications
| PRPF6 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| PRPF6 | |
| Synthetic peptides corresponding to PRPF6(PRP6 pre-mRNA processing factor 6 homolog (S. cerevisiae)) The peptide sequence was selected from the N terminal of PRPF6. Peptide sequence PGKRTVGDQMKKNQAADDDDEDLNDTNYDEFNGYAGSLFSSGPYEKDDEE. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Xenopus: 100%; Chicken: 100%; Canine: 100%; Guinea pig: 100%; Equine: 100%; Mouse: 100%; Rabbit: 100%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| Purified |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| androgen receptor N-terminal domain transactivating protein-1, ANT-1, bB152O15.1, C20orf14, chromosome 20 open reading frame 14, hPrp6, p102 U5 small nuclear ribonucleoprotein particle-binding protein, pre-mRNA-processing factor 6, Prp6, PRP6 homolog, PRP6 pre-mRNA processing factor 6 homolog (S. cerevisiae), PRP6 pre-mRNA processing factor 6 homolog (yeast), putative mitochondrial outer membrane protein import receptor, TOM, U5 snRNP-associated 102 kDa protein, U5-102 kDa protein, U5-102K | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 24148 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction