missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ PRP-h5 Reversed Phase Column

Delivers a selectivity difference from standard silica C18 stationary phases.

Brand:  Hamilton™ 79263

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12386479

  • £2205.00 / Each
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

Description

Description

  • Utilizes the PRP-3 as the base with a pentafluorinated modification, making it hydrophobic in nature
  • Functionality derived from a pentafluorinated polymer bead
  • Provides chromatographers the desired retention characteristics necessary for certain sample types
  • Maintains its performance characteristics longer than many silica-based C8 or C18 columns
  • Applications include organic compounds, pharmaceuticals, protein digests, tryptic digests and proteomics

Specifications

Specifications

Preparative Column
100 mm
5 μm
300 Å
5°C to 85°C
PSDVB (Polystyrene Divinylbenzene)
Reversed Phase
10 mm
5000 psi (345 bar)
1 to 13
1/Pk.
Pentafluorinated PSDVB
Reversed Phase
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers