Learn More
Hamilton™ PRP™-1 Guard Cartridge Kit
Stainless steel and PEEK guard columns help protect your investment
Brand: Hamilton™ 79317
Code : NEW
Additional Details : Weight : 0.16300kg
Description
- Guard columns remove particulate contaminants and highly adsorptive compounds from samples
- Modular and cartridge replacement is easy and tool free
- Holders are available in stainless steel and PEEK materials, both designs optimized to reduce dead volume and prevent extra column band broadening
- Holders for the cartridges are reusable indefinitely
Kit contains 1 holder and 2 cartridges
Recommended for use with virtually all solvents except high concentrations of Sodium Chloride.
Specifications
1 to 13 | |
8 mm | |
57 μL | |
3 mm | |
Virtually all solvents except high concentrations of sodium chloride |
10 μm | |
5000 psi | |
HPLC Guard Cartridge Kit | |
PEEK |