Learn More
Hamilton™ PRP™-1 Guard Cartridge Kit
Stainless steel and PEEK guard columns help protect your investment
Brand:  Hamilton™ 79447
Code : 16
Additional Details : Weight : 0.19100kg
 View more versions of this product
Product Code. 10705493
  • £330.00 / Each
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
Description
Description
  • Guard columns remove particulate contaminants and highly adsorptive compounds from samples
  • Modular and cartridge replacement is easy and tool free
  • Holders are available in stainless steel and PEEK materials, both designs optimized to reduce dead volume and prevent extra column band broadening
  • Holders for the cartridges are reusable indefinitely
Kit contains 1 holder and 2 cartridges
Recommended for use with virtually all solvents except high concentrations of Sodium Chloride.
Specifications
Specifications
1 to 13
20 mm
80 μL
2.3 mm
Virtually all solvents except high concentrations of sodium chloride
10 μm
5000 psi
HPLC Guard Cartridge Kit
Stainless Steel