missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Protino Ni-IDA 2000 kit for His-tag Protein Purification in Maxi format
Click to view available options
Unit Size:
Pack of 25
Pack of 5
Description
Protino™ Ni-IDA, the purer alternative for higher yields of His-tag proteins
- Chelating group IDA, three protein binding sites
- High protein yield/recovery even from diluted samples
- High protein concentration
- High binding specificity, less unspecific binding of contaminating proteins compared to other common IMAC matrices
- Dry resin, storage at room temperature
- Ready to use pre-packed columns filled with dry Protino™ Ni-IDA resin
- Buffers included
- Dry bulk matrix
- Rigid support silica matrix, compatible with high pressure and high flow rates, does not shrink or swell - highest flexibility
- Batch procedure, gravity flow column chromatography, FPLC™ applications
- Empty Protino™ columns available for use with Protino™ Ni-IDA resin
- Empty 96 well receiver plates available for use with Protino™ Ni-IDA resin
- Ready to use pre-packed 96 well plates filled with dry Protino™ Ni-IDA resin
- Buffers included For purification of polyhistidine-tagged proteins, batch binding, gravity flow column chromatography, MPLC/FPLC™ applications.
Specifications
Specifications
| Bed Volume | 1000 μL |
| Binding Capacity | 10mg |
| Binding Property | Batch |
| Kit Contents | Buffers |
| Matrix | Macroporous Silica |
| For Use With (Application) | For Purification of Polyhistidine-Tagged Proteins |
| Type | Ni-TED Protein Purification Kit |
| Ligand Type | IDA (iminodiacetic acid) |
| Sufficient For | 25 Reactions |
Safety and Handling
Product Identifier
- Protino Ni-IDA 2000 packed columns (25)
- Danger
- Reproductive toxicity Category 1B
- H360D-May damage the unborn child.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- MIXTURE LIST-Contains : imidazole, monosodium phosphate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction