missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Protino Ni-IDA 150 kit for His-tag Protein Purification in Mini format
Description
Protino™ Ni-IDA, the purer alternative for higher yields of His-tag proteins
- Chelating group IDA, three protein binding sites
- High protein yield/recovery even from diluted samples
- High protein concentration
- High binding specificity, less unspecific binding of contaminating proteins compared to other common IMAC matrices
- Dry resin, storage at room temperature
- Ready to use pre-packed columns filled with dry Protino™ Ni-IDA resin
- Buffers included
- Dry bulk matrix
- Rigid support silica matrix, compatible with high pressure and high flow rates, does not shrink or swell - highest flexibility
- Batch procedure, gravity flow column chromatography, FPLC™ applications
- Empty Protino™ columns available for use with Protino™ Ni-IDA resin
- Empty 96 well receiver plates available for use with Protino™ Ni-IDA resin
- Ready to use pre-packed 96 well plates filled with dry Protino™ Ni-IDA resin
- Buffers included For purification of polyhistidine-tagged proteins, batch binding, gravity flow column chromatography, MPLC/FPLC™ applications.
Specifications
Specifications
| Bed Volume | 80 μL |
| Binding Capacity | 0.8mg |
| Kit Contents | Buffers |
| Matrix | Macroporous Silica |
| Quantity | 50 Reactions |
| For Use With (Application) | For Purification of Polyhistidine-Tagged Proteins |
| Type | Ni-TED Protein Purification Kit |
| Ligand Type | IDA (iminodiacetic acid) |
Safety and Handling
Product Identifier
- Protino Ni-IDA 150 packed columns (50)
- Danger
- Reproductive toxicity Category 1B
- H360D-May damage the unborn child.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- MIXTURE LIST-Contain: imidazole, monosodium phosphate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?