missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Protino Ni-TED 2000 kit for His-tag Protein Purification in Maxi format
Click to view available options
:
No. of Reactions: 5
Description
Protino™ NI-TED, the purer alternative for His-tag protein purification
- Chelating group TED, one protein binding site
- High stability against reducing/chelating agents
- Low metal leaching
- High protein purity
- High binding specificity, less unspecific binding of contaminating proteins compared to other common IMAC matrices
- Dry resin, storage at room temperature
- Ready to use pre-packed columns filled with dry Protino™ Ni-TED resin
- Buffers included
- Dry bulk matrix
- Rigid support silica matrix, compatible with high pressure and high flow rates, does not shrink well - highest flexibility
- Batch procedure, gravity flow column chromatography, FPLC™ applications
- Empty Protino™ columns available for use with Protino™ Ni-TED resin
- Empty 96 well receiver plates available for use with Protino™ Ni-TED resin For purification of polyhistidine-tagged proteins; batch binding, gravity flow column chromatography, MPLC/FPLC™ applications.
Specifications
Specifications
| Bed Volume | 1000 μL |
| Binding Capacity | 5mg |
| Binding Property | Batch |
| Kit Contents | Buffers |
| Matrix | Macroporous Silica |
| Quantity | 5 Reactions |
| For Use With (Application) | For Purification of Polyhistidine-Tagged Proteins, Batch Binding, Gravity Flow Column Chromatography, MPLC/FPLC Applications |
| Type | Ni-TED Protein Purification Kit |
| Ligand Type | TED (tris-carboxymethyl ethylene diamine) |
Safety and Handling
Hazard Category
- Respiratory sensitiser Category 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction