missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Protino Ni-TED 1000 kit for His-tag Protein Purification in Midi format
Click to view available options
:
50 prep kit of packed columns for the purification of His-Tag proteins, buffer LEW, elution buffer, protocol Macherey-Nagel
Description
Protino™ NI-TED, the purer alternative for His-tag protein purification
- Chelating group TED, one protein binding site
- High stability against reducing/chelating agents
- Low metal leaching
- High protein purity
- High binding specificity, less unspecific binding of contaminating proteins compared to other common IMAC matrices
- Dry resin, storage at room temperature
Protino™ NI-TED packed columns
- Ready to use pre-packed columns filled with dry Protino™ Ni-TED resin
- Buffers included
Protino™ NI-TED resin
- Dry bulk matrix
- Rigid support silica matrix, compatible with high pressure and high flow rates, does not shrink well - highest flexibility
- Batch procedure, gravity flow column chromatography, FPLC™ applications
- Empty Protino™ columns available for use with Protino™ Ni-TED resin
- Empty 96 well receiver plates available for use with Protino™ Ni-TED resin
For purification of polyhistidine-tagged proteins; batch binding, gravity flow column chromatography, MPLC/FPLC™ applications.
Specifications
Specifications
| Bed Volume | 500 μL |
| Binding Capacity | 2.5mg |
| Kit Contents | Protino™ Ni-TED 1000 Packed Columns, buffers |
| Matrix | Macroporous silica |
| Content And Storage | Room Temperature |
| Quantity | 1 Kit |
| For Use With (Application) | Purification of polyhistidine-tagged proteins |
| Type | Ni-TED Protein Purification Kit |
| Ligand Type | TED (tris-carboxymethyl ethylene diamine) |
Safety and Handling
Product Identifier
- Protino Ni-TED 1000 packed columns (50)
- Danger
- Reproductive toxicity Category 1B
- H360D-May damage the unborn child.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- MIXTURE LIST-Contains : imidazole, monosodium phosphate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction