missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Protino Ni-NTA Agarose for His-tag Protein Purification

Product Code. 12718702
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 12718702

Brand: Macherey-Nagel™ 745400.100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Method of choice for His-tag protein purification with best performance.

Specifications

Binding Capacity <50 mg/mL settled agarose
Capacity (Metric) 100 mL
Binding Property Batch
Matrix 6 % Beaded Agarose (Cross-Linked), Precharged With Ni2+
Content And Storage 4°C to 8°C
Quantity 100 mL
Type Agarose
Ligand Type NTA (nitrilotriacetic acid)
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : ethanol (diluted < 20 %) (denatured with1%IPA/1%MEK, acc.2016/1867/EU), resin suspension with Ni in
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.