missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Protino Ni-IDA Resin for His-tag Protein Purification
Click to view available options
Quantity:
30 g Dry Silica Bulk Resin
5 g Dry Silica Bulk Resin
Unit Size:
30g
5g
Description
Protino™ Ni-IDA, the purer alternative for higher yields of His-tag proteins
- Chelating group IDA, three protein binding sites
- High protein yield/recovery even from diluted samples
- High protein concentration
- High binding specificity, less unspecific binding of contaminating proteins compared to other common IMAC matrices
- Dry resin, storage at room temperature
- Ready to use pre-packed columns filled with dry Protino™ Ni-IDA resin
- Buffers included
- Dry bulk matrix
- Rigid support silica matrix, compatible with high pressure and high flow rates, does not shrink or swell - highest flexibility
- Batch procedure, gravity flow column chromatography, FPLC™ applications
- Empty Protino™ columns available for use with Protino™ Ni-IDA resin
- Empty 96 well receiver plates available for use with Protino™ Ni-IDA resin
- Ready to use pre-packed 96 well plates filled with dry Protino™ Ni-IDA resin
- Buffers included For purification of polyhistidine-tagged proteins, batch binding, gravity flow column chromatography, MPLC/FPLC™ applications.
Specifications
Specifications
| Binding Capacity | 20mg/g |
| Binding Property | Batch |
| Flow Rate | 0.5 to 1.0 mL/min./cm2 |
| Kit Contents | Buffers |
| Matrix | Macroporous Silica |
| Max. Operating Pressure | 145 psi |
| Quantity | 5 g Dry Silica Bulk Resin |
| For Use With (Application) | For Purification of Polyhistidine-Tagged Proteins, Batch Binding, Gravity Flow Column Chromatography, MPLC/FPLC Applications |
| Type | Resin |
| Ligand Type | IDA (iminodiacetic acid) |
Safety and Handling
Signal Word
- Danger
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1A
- LONG-TERM AQUATIC HAZARD Chronic 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P391-Collect spillage.
- P405-Store locked up.
- MIXTURE LIST-Contains : resin with Ni in nickel(II) complexes
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction