missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Protino Glutathione Agarose 4B for GST-tag Protein Purification

Product Code. 11912302
Click to view available options
Quantity:
10 mL
100 mL
Unit Size:
100mL
10mL
This item is not returnable. View return policy
Do not autoclave the gel.

Product Code. 11912302

Brand: Macherey-Nagel™ 745500.100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Do not autoclave the gel.

Best choice for cost effective GST-tag protein purification.

Protino™ Glutathione Agarose 4B is an affinity chromatography medium that enables fast, single-step purification of glutathione-S-transferase (GST) fusion proteins and other glutathione-binding proteins. Purification of fusion proteins from a whole cell lysate is based on the strong affinity of the GST moiety for glutathione, which is immobilized on agarose beads.

  • Highest performance–Equivalent to Glutathione Sepharose™ 4B / GSTrap™ 4B columns
  • Simply replace your current products—No optimization or protocol change necessary
  • Universal usage—Suitable for small proteins, large protein complexes, proteins with low expression rates universal use
  • Highest flexibility of applications—For batch binding, gravity-flow column chromatography, MPLC/FPLC™

Specifications

Bed Volume 100 mL
Binding Capacity 8mg/mL
Concentration 0.1 M acetate, pH 4, 0.1 M NaOH, 70% ethanol, or 6 M guanidine hydrochloride
Flow Rate 250 cm/hr.
Material Settled beaded agarose
Matrix 4% beaded agarose
Content And Storage 4°C to 8°C for up to 3 years
Particle Size 90 μm
Quantity 100 mL
For Use With (Application) Purification of GST-tagged proteins using affinity chromatography
Type Agarose
Sample Volume 0.2 to 1.0 mL/min. (with 1 mL bed volume/column with 6.6 mm I.D.), 0.5 to 5.0 mL/min. (with 10 mL bed volume/column with 16 mm I.D.)
Ligand Type Chelating ligand: Glutathione, linked via sulfur atom
Show More Show Less
compliance-icons
Product Identifier
  • Protino Glutathione Agarose 4B
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Supplemental information
  • MIXTURE LIST-Contains : ethanol (diluted < 20 %) (denatured with1%IPA/1%MEK, acc.2016/1867/EU)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.