missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Protino 96-well Ni-NTA kit for His-tag Protein Purification

Product Code. 12860296
Change view
Click to view available options
Bed Volume:
50 μL
Unit Size:
384 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Volume unitSize
12860296 50 μL 384 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12860296 Supplier Macherey-Nagel™ Supplier No. 745425.4

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Method of choice for His-tag protein purification with best performance in 96‐well plate format.

Specifications

Bed Volume 50 μL
Binding Capacity 2 mg/well with 50μL of Settled Agarose
Matrix 6% Beaded Agarose (Cross-Linked), Precharged With Ni2+
Content And Storage 4°C to 8°C
Type 96-well Protein Purification Plate
Ligand Type NTA (nitrilotriacetic acid)
compliance-icons compliance-icons
Hazard Category
  • Respiratory sensitiser Category 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.