missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel Bioanalysis™ Protino™ 96 Ni-Nta

Product Code. 12850296
Change view
Click to view available options
Quantity:
384 Preps
Unit Size:
96 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12850296 384 Preps 96 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12850296 Supplier Macherey Nagel Bioanalysis™ Supplier No. 745425.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Method of choice for His-tag protein purification with best performance in 96‐well plate format

Protino™ Ni-NTA agarose, Protino™ purification plate, MN wash plate 1 x 96 preps Macherey-Nagel

Specifications

Bed Volume Variable (50 μL recommended)
Content And Storage 4°C to 8°C
Sample Type Protein solution
Particle Size 45 to 165 μm
Quantity 384 Preps
For Use With (Application) Purification of His-tag proteins
Target Protein
Format 96-well Plate
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.