missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Protien G Sepharose High Performance
Shop All Cytiva Products
Click to view available options
Quantity:
200 mL
Description
- Ligand: Recombinant protein G lacking albumin-binding region
- Matrix: Highly cross-linked agarose, 6%
- Chemical stability: All common used buffers
- Storage: 4 to 8°C, 20% Ethanol
Specifications
Specifications
| Binding Capacity | >1 mg lgG/column |
| Capacity (Metric) | 200 mL |
| Matrix | Highly cross-linked agarose, 6% |
| pH Range | 3 to 9 |
| Content And Storage | 4 to 8 °C, 20% Ethanol |
| Particle Size | ∼34 μm |
| Quantity | 200 mL |
| Ligand Type | Recombinant protein G lacking albumin-binding region |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur