missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Prothrombin Fragment 1/2 Monoclonal Antibody (D3), Invitrogen™

Mouse Monoclonal Antibody
£161.00 - £550.00
Specifications
| Antigen | Prothrombin Fragment 1/2 |
|---|---|
| Clone | D3 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17989222
![]() |
Invitrogen™
MA550945 |
200 μL |
£550.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17960733
|
Invitrogen™
MA550946 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Prothrombin Fragment 1/2 is a protein fragment that is produced by the cleavage of prothrombin, a blood clotting protein. It is a small peptide consisting of the first half of the prothrombin molecule and is involved in the formation of blood clots. Prothrombin Fragment 1/2 is used as a biomarker for the diagnosis and monitoring of various blood clotting disorders.Specifications
| Prothrombin Fragment 1/2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| prothrombin fragment 1.2; Prothrombin fragment 1+2; prothrombin fragment F1.2; Prothrombin fragment F1+2 | |
| F2 | |
| IgG2a λ | |
| Protein A/G | |
| F2 |
| D3 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Pig | |
| P00734, Q19AZ8 | |
| 100144442, 2147 | |
| Recombinant Human F1+2 protein, Ala44-Arg327 (Accession #P00734). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title