missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Prothrombin Fragment 1/2 Monoclonal Antibody (D3), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA550945
This item is not returnable.
View return policy
Description
Prothrombin Fragment 1/2 is a protein fragment that is produced by the cleavage of prothrombin, a blood clotting protein. It is a small peptide consisting of the first half of the prothrombin molecule and is involved in the formation of blood clots. Prothrombin Fragment 1/2 is used as a biomarker for the diagnosis and monitoring of various blood clotting disorders.
Specifications
| Prothrombin Fragment 1/2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P00734, Q19AZ8 | |
| F2 | |
| Recombinant Human F1+2 protein, Ala44-Arg327 (Accession #P00734). | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG2a λ |
| Immunohistochemistry (Paraffin), Western Blot | |
| D3 | |
| Unconjugated | |
| F2 | |
| prothrombin fragment 1.2; Prothrombin fragment 1+2; prothrombin fragment F1.2; Prothrombin fragment F1+2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100144442, 2147 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction