missing translation for 'onlineSavingsMsg'
Learn More

Beckman Coulter Proteinase K (GeneFind V3 Kit)

Product Code. 31723104
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31723104 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Refer to SDS
Product Code. 31723104 Supplier Beckman Coulter Supplier No. C34821

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Refer to SDS

Proteinase K enzyme used to remove proteins from samples.

  • Proteinase K is a proteolytic enzyme formulated for effective protein removal during sample preparation workflows.
  • Helps break down proteins to support cleaner sample extracts and improved downstream processing.
  • Supplied as Proteinase K for use with the GeneFind V3 Kit and compatible laboratory protocols.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mL
Product Type Proteinase K
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.