missing translation for 'onlineSavingsMsg'
Learn More

Beckman Coulter Proteinase K

Product Code. 31723146
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31723146 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Refer to SDS
Product Code. 31723146 Supplier Beckman Coulter Supplier No. C34827

Please to purchase this item. Need a web account? Register with us today!

Refer to SDS

Proteinase K enzyme used to remove proteins from samples.

  • Proteinase K is a proteolytic enzyme formulated for effective protein removal during sample preparation workflows.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mL
Product Type Proteinase K
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.