missing translation for 'onlineSavingsMsg'
Learn More

Melford Proteinase K

Product Code. 30018717
Click to view available options
Quantity:
25 mg
100 mg
500 mg
1 g
Unit Size:
100mg
1g
25mg
500mg
This item is not returnable. View return policy

Product Code. 30018717

Brand: Melford P502201.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

From Tritirachium album.

From Tritirachium album. Serine protease for proteolytic inactivation of nucleases during isolation of DNA and RNA.

Description

Proteinase K is a highly reactive serine protease that displays an ability to digest native proteins, thereby inactivating enzymes such as DNase and RNase without recourse to a denaturation process. It is the most powerful proteinase among all proteinases characterized so far. It cleaves at the peptide bond adjacent to the carboxylic acid group of aliphatic, aromatic, or hydrophobic amino acids.

Recombinant Proteinase K is used in the isolation or preparation of high molecular weight nucleic acids. It is highly pure has a higher specific activity and is more stable at room temperature when compared to native Proteinase K.

Recombinant Proteinase K is expressed from yeast Pichia pastoris with a cloned gene encoding Engyodontium album (Tritirachium album) endolytic protease.

Applications

  • Recombinant Proteinase K is extremely effective on native proteins and can therefore be used to rapidly inactivate endogenous nucleases such as RNases and DNases. This property makes Proteinase K particularly suitable for the isolation of native RNA and DNA from tissues or cell lines.
  • Used for the analysis of membrane structures by modifying proteins and glycoproteins on cell surfaces.
  • Particularly well suited for isolating nucleic acids for amplification reactions.
  • Promotes cell lysis by activating a bacterial autolytic factor.
  • Used to remove cellular debris during the preparation of colony lifts, and to treat tissue sections to ensure efficient probe infiltration during in situ hybridization.

Reconstitution Recommendations:

For stock solution preparation, we suggest reconstituting the lyophilized powder in a buffer (20 mM Tris, 1 mM CaCl2, 2% Glycerol, pH 7.4).

Calcium is important for thermo-stability of Proteinase K and pH value is critical for high solubility.

The use of cryoprotectant glycerol to a final concentration of 50% in stock solution is to prevent freeze/thaw damage. Proteinase K retained 99% of activity after 12 freeze/thaw cycles in stock solution. Avoid repeated freezing and thawing to prevent precipitation of the protein if glycerol is not added.

Proteinase K solution remains active over a broad pH range (4.0-12.5, often used in range of 7.5-9.0) and over a broad temperature range of 25°C to 75°C. Solution remains sterile and stable for at least 12 months at 4°C.

Caution: Proteinase K Recombinant Lyophilized Powder is not sterile. Stock solution can be prepared as a 20-40 mg/mL in 20 mM Tris-HCl buffer, sterilized using a 0.22 μm filter. Stock solution can be stored in 50% glycerol in aliquots at a wide temperature range of 24°C to -80°C.

For sterile filtration, we recommend using PES pr PVDF membranes with low protein binding.

Composition (NO ADDITIVES):

  • ≥90% Protein Proteinase K, biuret test.
  • Excipients: Sucrose 8%
  • DNA and RNA free
  • Endotoxin Free

Specifications:

  • Stable and active over a wide pH range of 4 to 12, Maximal Activity is in pH range of 7.5 to 12.0
  • Active with SDS, urea and EDTA. The most active temperature is 65°C.
  • Inactivated by diisopropyl fluorophosphates (DFP) and phenyl methane sulfonyl fluoride (PMSF).
  • Purification: Nickel affinity chromatography followed by ion-exchange chromatography and dialysis.
  • EC : 3.4.21.64

Specifications

CAS 39450-01-6
Quantity 1 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.