missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein A Antibody, R&D Systems™

Rat Monoclonal Antibody
£201.00 - £463.00
Specifications
| Antigen | Protein A |
|---|---|
| Clone | 946412 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18740607
![]() |
R&D Systems
MAB9976-SP |
25 μg |
£201.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18792017
![]() |
R&D Systems
MAB9976-100 |
100 μg |
£463.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Protein A Monoclonal antibody specifically detects Protein A in Broad Species Reactivity samples. It is validated for ELISASpecifications
| Protein A | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 3919448 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 946412 | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Multi-species | |
| Immunoglobulin G binding protein A, PROA, Protein-A, SPA, Staphylococcal protein A | |
| Recombinant Protein A | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title