missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein A Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB9976-100
This item is not returnable.
View return policy
Description
Protein A Monoclonal antibody specifically detects Protein A in Broad Species Reactivity samples. It is validated for ELISA
Specifications
| Protein A | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3919448 | |
| Multi-species | |
| Purified |
| ELISA | |
| 946412 | |
| ELISA | |
| Immunoglobulin G binding protein A, PROA, Protein-A, SPA, Staphylococcal protein A | |
| Recombinant Protein A | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction