missing translation for 'onlineSavingsMsg'
Learn More

3M™ Protective Coveralls

Product Code. 15217449
Click to view available options
:
Size: Large; Color: Grey
This item is not returnable. View return policy

Product Code. 15217449

Brand: 3M™ 7000089689

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

With a breathable back panel to improve air ventilation and reduce heat build-up. 3M™ Protective Coveralls are manufactured from a high quality laminated material.

  • Available in a range of sizes
  • Extra comfort for extended wear
  • Provides barrier protection against light liquid splashes and hazardous dust
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protective Coveralls
Size Large
Material Polyethylene
Color Gray
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Closure Type Zipper
Resistance Liquid
Back Style Large Zip
Certifications/Compliance CE Category III
Description Enlarged Zip, Finger Loops, a Double-Closing Storm Flap System and Chin Flap; Low Linting Material
For Use With (Application) Chemical Manufacturing, Oil Spills and Clean Up Hazardous Waste Remediation Decontamination Tank Cleaning
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.