missing translation for 'onlineSavingsMsg'
Learn More

3M™ Protective Coverall 4540+, Type 5/6

Catalog Number 15415613
Click to view available options
Size:
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

With premium dual material technology and offering CE Category III, Type 5/6 protection, the 3M™ Protective Coverall 4540+ is ideal for applications where both high levels of protective performance and comfort desired.

  • Type 5/6 highly versatile protective coverall designed to offer an excellent barrier to particles and limited liquid splash and also features a breathable SMMMS back panel
  • Excellent barrier against dry particles and certain liquid chemical splash (CE Type 5/6)
  • Soft and light, ultra low-linting laminated microporous material that drapes and hangs well over the body facilitating ease of movement
  • Back panel made of breathable SMMMS material for improved air circulation and reduced heat stress
  • 2-way zipper with fully sealable storm flap for added on-off convenience and extra protection
  • 3-panel hood design for enhanced compatability with complimentary PPE
  • Knitted cuffs and elasticated waist and ankles for increased comfort and freedom of movement
  • Anti-static treated
  • Low linting
Product Type Cleanroom Coveralls
Size 4X-Large
Material SMS
Color White, Blue
Disposable Single-use
Waist Style Elastic
Cuff Style Knit
Hood Type 3-panel
Closure Type 2-way Zipper
Seam Type Serged, Bound
Sterility Non-sterile
Certifications/Compliance EN ISO 13982-1:2004 Type 5 Hazardous Dusts, EN 13034:2005+A1:2009 Type 6 Liquid Splash, CE Category III, EN 1149-5 - Electrostatic Properties, EN 1073-2 Particulate Radioactive Contamination
For Use With (Application) Controlled Environments
Style Enhanced Gusset
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'