missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome 19S 10B Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£343.00 - £557.00
Specifications
| Antigen | Proteasome 19S 10B |
|---|---|
| Dilution | Western Blot 0.04 to 0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18335226
|
Novus Biologicals
NBP3-17019-25UL |
25 μg |
£343.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18327876
|
Novus Biologicals
NBP3-17019-100UL |
100 μg |
£557.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Proteasome 19S 10B Polyclonal antibody specifically detects Proteasome 19S 10B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Proteasome 19S 10B | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Neuroscience, Ubiquitin Proteasome Pathway | |
| PBS, pH 7.2, 40% glycerol | |
| 5708 | |
| IgG | |
| Affinity purified |
| Western Blot 0.04 to 0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 26S proteasome regulatory subunit RPN1, 26S proteasome regulatory subunit S2, MGC14274,55.11 protein, P97, proteasome (prosome, macropain) 26S subunit, non-ATPase, 2, Protein 55.11, Rpn1, S226S proteasome non-ATPase regulatory subunit 2, TNFR-associated protein 2, TRAP226S proteasome subunit p97, tumor necrosis factor receptor-associated protein 2, Tumor necrosis factor type 1 receptor-associated protein 2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: EYEDLTEIMSNVQLNSNFLALARELDIMEPKVPDDIYKTHLENNRFGGSGSQVDSARMNLASSFVNGFVNAAFGQDKLLTDDGNKWLYKNKD | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel