missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Protease QF
Click to view available options
Quantity:
180 μL
Unit Size:
180µL
Description
Protease QF, 180 μL, is an enzyme in aqueous buffered solution and an individual component of the ViewRNA™ ISH Tissue Assay Kits, 2-Plex, 24 assays.
Specifications
Specifications
| Content And Storage | Store at 2°C to 8°C IMPORTANT! Do not freeze. |
| Form | Liquid |
| Quantity | 180 μL |
| Product Type | Protease |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction