missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Propidium Iodide Ready Flow™ Reagent

Product Code. 15703019
Change view
Click to view available options
Quantity:
120 assays
Unit Size:
Pack of 2
Product Code. Quantity unitSize
15703019 120 assays Pack of 2
1 options
This item is not returnable. View return policy

Product Code. 15703019

Brand: Invitrogen™ R37169

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Propidium Iodide Ready Flow™ Reagent is a bright, easy-to-use nucleic acid stain for distinguishing dead cells from live cells in flow cytometry assays.

Propidium Iodide Ready Flow™ Reagent is a bright, easy-to-use nucleic acid stain for distinguishing dead cells from live cells in flow cytometry assays. This dye has excitation/emission maxima of 535/617 nm.

• Stable at room temperature
• Convenient, ready-to-use format—no need to dilute, weigh. or pipette
• Rapid staining of dead cells without wash steps

Propidium Iodide Ready Flow Reagent is a commonly used dead-cell stain that emits red fluorescence when bound to DNA. Add 2 drops from the dropper bottle directly into 1 x 106 cells, incubate, and analyze.

Propidium iodide is membrane impermeant and therefore excluded from viable cells. It binds to DNA by intercalating between the bases, thereby binding both DNA and RNA. Propidium iodide is commonly excited with the 488 nm laser, but others such as the 532 and 561 nm lasers are also suited for PI excitation.

TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Cell-Impermeant
Color Red
Content And Storage 2 dropper bottles, store (at room temperature)
Detection Method Fluorescence
Product Type Fluorescent Stain
Product Line Ready Flow
Quantity 120 assays
Reagent Type Nucleic Acid Stain
Sub Cellular Localization Nucleus
Excitation/Emission 535/617
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.