missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Propidium Iodide

Product Code. 15541957
Change view
Click to view available options
Quantity:
1.5 mL
Unit Size:
1.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15541957 1.5 mL 1.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15541957 Supplier Invitrogen™ Supplier No. BMS500PI
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Propodium Iodide

Standard reagent used for assessing cell viability and exclusion of non-viable cells in flow cytometry. PI binds to double-stranded DNA, but is excluded from cells with intact plasma membranes.

  • PI should be analyzed in the PE channel when used as a counterstain for Annexin V FITC
  • 7-AAD can be used in place of PI when using Annexin V PE
  • The advantage of 7-AAD over PI is that there is minimal spectral overlap between these emissions
  • Choose Propidium Iodide as a counterstain with Annexin V apoptosis assays
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Propidium Iodide
Dye Type Cell-Impermeant
Emission 535/617
Product Line eBioscience
Quantity 1.5 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.