Learn More
Thermo Scientific™ ProPac™ IMAC-10 HPLC Columns
Resolve target proteins using a single column in a high-resolution gradient run with ProPac IMAC-10 LC Columns.
£150.66 - £1953.32
Specifications
Max. Pressure | 3000 psi (207 bar) |
---|---|
Particle Size | 10 μm |
pH | 2 to 12 |
Pore Size | Nonporous |
Temperature | 60°C |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Column Format | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Column Format | Price | Quantity & Availability | ||
11331823
|
Thermo Scientific™
063272 |
10 μm | 2 mm | 50 mm | Analytical Column |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11301843
|
Thermo Scientific™
063274 |
10 μm | 2 mm | 250 mm | Analytical Column |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11311843
|
Thermo Scientific™
063276 |
10 μm | 4 mm | 50 mm | Analytical Column |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11321843
|
Thermo Scientific™
063278 |
10 μm | 4 mm | 250 mm | Analytical Column |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11331843
|
Thermo Scientific™
063280 |
10 μm | 9 mm | 250 mm | Analytical Column |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11381853
|
Thermo Scientific™
063615 |
10 μm | 9 mm | 50 mm | Analytical Column |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11391853
|
Thermo Scientific™
063617 |
10 μm | 1 mm | 50 mm | Analytical Column |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11331863
|
Thermo Scientific™
063667 |
10 μm | 4 mm | 50 mm | Loading Column |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11311873
|
Thermo Scientific™
063710 |
10 μm | 9 mm | 50 mm | Loading Column |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Resolve target proteins using a single column in a high-resolution gradient run with Thermo Scientific™ ProPac™ IMAC-10 LC Columns. These analytical and semi-preparative HPLC columns provides high-purity separations of metal-binding proteins and high loading capacity for protein purification applications. The use of small, nonporous resin particles results in narrow, high-efficiency peaks, with minimal band broadening due to dispersion. Use loading columns to charge the ProPac IMAC-10 LC columns but avoid contact of the metal solution with the system.
Tailored Specificity for IMAC Separation of Proteins- Retention control by imidazole concentration or pH gradient
- Stable between pH 2 to 12, compatible with common protein purification reagents (nonionic detergents, reducing agents)
- Effective under native or denaturing HPLC conditions
- Wide range of metal-specific applications, including the separation of His-tagged protein aggregation variants, prion peptide analysis and separation of monoclonal antibodies
- Shipped free from any metal and ready to be charged with the metal of your choice.
- Reusable columns with metal tailored specificity
- Resolve target proteins using a single column in a high resolution gradient run
- High purity separations of metal-binding proteins
Specifications
3000 psi (207 bar) | |
2 to 12 | |
60°C | |
Spherical, Nonporous Polymer | |
Affinity Separation |
10 μm | |
Nonporous | |
Lysozyne Poly(IDA) Grafts | |
Separation of Metal-Binding Proteins |