missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ProPac™ IMAC-10 HPLC Columns

Product Code. 11311843
Change view
Click to view available options
Length (Metric):
50 mm
250 mm
Diameter (Metric):
1 mm
2 mm
4 mm
9 mm
Column Format:
Analytical Column
Loading Column
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Column Format unitSize
11311843 50 mm 4 mm Analytical Column Each
11331823 50 mm 2 mm Analytical Column Each
11301843 250 mm 2 mm Analytical Column Each
11321843 250 mm 4 mm Analytical Column Each
11331843 250 mm 9 mm Analytical Column Each
11381853 50 mm 9 mm Analytical Column Each
11391853 50 mm 1 mm Analytical Column Each
11331863 50 mm 4 mm Loading Column Each
11311873 50 mm 9 mm Loading Column Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 11311843 Supplier Thermo Scientific™ Supplier No. 063276
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Resolve target proteins using a single column in a high-resolution gradient run with ProPac IMAC-10 LC Columns.

Resolve target proteins using a single column in a high-resolution gradient run with Thermo Scientific™ ProPac™ IMAC-10 LC Columns. These analytical and semi-preparative HPLC columns provides high-purity separations of metal-binding proteins and high loading capacity for protein purification applications. The use of small, nonporous resin particles results in narrow, high-efficiency peaks, with minimal band broadening due to dispersion. Use loading columns to charge the ProPac IMAC-10 LC columns but avoid contact of the metal solution with the system.

Tailored Specificity for IMAC Separation of Proteins
  • Retention control by imidazole concentration or pH gradient
  • Stable between pH 2 to 12, compatible with common protein purification reagents (nonionic detergents, reducing agents)
  • Effective under native or denaturing HPLC conditions
  • Wide range of metal-specific applications, including the separation of His-tagged protein aggregation variants, prion peptide analysis and separation of monoclonal antibodies
  • Shipped free from any metal and ready to be charged with the metal of your choice.
  • Reusable columns with metal tailored specificity
  • Resolve target proteins using a single column in a high resolution gradient run
  • High purity separations of metal-binding proteins
Includes: 10μm, nonporous, polymeric beads coated with a hydrophilic layer, then grafted with poly(IDA) chains.
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 4 mm
Flow Rate 1 mL/min
For Use With UltiMate 3000 HPLC System
Length (Metric) 50 mm
Max. Pressure 3000 psi (207 bar)
Mobile Phase Compatibility pH 2 to 12, compatible with most reagents used in IMAC protein purifications, including EtOH, Urea, NaCl, non-ionic detergents, glycerol, acetic acid, guanidine, HCl
Particle Size 10 μm
pH 2 to 12
Pore Size Nonporous
Quantity Each
Temperature 60°C
Stationary Phase Lysozyne Poly(IDA) Grafts
Product Line ProPac
Packing Material Spherical, Nonporous Polymer
Column Type Separation of Metal-Binding Proteins
Phase Affinity Separation
Type Analytical Column
Technique Bio-LC
Green Features FSC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.