missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PROP1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 1 publication
£136.00 - £332.00
Specifications
| Antigen | PROP1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Peptide ELISA Detection limit 1:32000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18543472
|
Novus Biologicals
NBP1-44054-25ug |
25 ug |
£136.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18206191
|
Novus Biologicals
NBP1-44054 |
0.1 mg |
£332.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PROP1 Polyclonal specifically detects PROP1 in Human samples. It is validated for Peptide ELISA.Specifications
| PROP1 | |
| Peptide ELISA Detection limit 1:32000 | |
| Polyclonal | |
| Goat | |
| Human | |
| NP_006252.2 | |
| 5626 | |
| Peptide with sequence AERRRQAEKPKKGR-C corresponding to N-Terminus according to NP_006252.2. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| ELISA | |
| Unconjugated | |
| RUO | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CPHD2, homeobox protein prophet of Pit-1, Pituitary-specific homeodomain factor, PROP paired-like homeobox 1, PROP-1, prophet of Pit1, paired-like homeodomain transcription factor | |
| PROP1 | |
| IgG | |
| Affinity Purified | |
| This antibody is specific to the N-Terminus. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title