missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PROP1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-44054-25ug
This item is not returnable.
View return policy
Description
PROP1 Polyclonal specifically detects PROP1 in Human samples. It is validated for Peptide ELISA.
Specifications
| PROP1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CPHD2, homeobox protein prophet of Pit-1, Pituitary-specific homeodomain factor, PROP paired-like homeobox 1, PROP-1, prophet of Pit1, paired-like homeodomain transcription factor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5626 | |
| Human | |
| IgG |
| ELISA | |
| 0.5 mg/mL | |
| Peptide ELISA Detection limit 1:32000 | |
| NP_006252.2 | |
| PROP1 | |
| Peptide with sequence AERRRQAEKPKKGR-C corresponding to N-Terminus according to NP_006252.2. | |
| 25 ug | |
| Primary | |
| This antibody is specific to the N-Terminus. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction