missing translation for 'onlineSavingsMsg'
Learn More

Prohibitin 2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£177.00 - £479.00

Specifications

Antigen Prohibitin 2
Dilution Western Blot 1:1000, Immunohistochemistry 1:50 - 1: 100, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18179717
View Documents
Novus Biologicals
NBP2-50550
400 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
18610837
View Documents
Novus Biologicals
NBP2-50550-100ul
100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Prohibitin 2 Polyclonal specifically detects Prohibitin 2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Prohibitin 2
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS with 0.09% Sodium Azide
11331
KLH conjugated synthetic peptide between 225-255 amino acids from human PHB2.
Primary
Western Blot 1:1000, Immunohistochemistry 1:50 - 1: 100, Immunohistochemistry-Paraffin
Polyclonal
Rabbit
Human, Mouse
BAP, Bap37, BCAP37, B-cell associated protein, D-prohibitin, MGC117268, p22, PNAS-141, prohibitin 2, prohibitin-2, REAB-cell receptor-associated protein BAP37, Repressor of estrogen receptor activity
PHB2
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.