missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prohibitin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-50550
This item is not returnable.
View return policy
Description
Prohibitin 2 Polyclonal specifically detects Prohibitin 2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Prohibitin 2 | |
| Polyclonal | |
| Western Blot 1:1000, Immunohistochemistry 1:50 - 1: 100, Immunohistochemistry-Paraffin | |
| BAP, Bap37, BCAP37, B-cell associated protein, D-prohibitin, MGC117268, p22, PNAS-141, prohibitin 2, prohibitin-2, REAB-cell receptor-associated protein BAP37, Repressor of estrogen receptor activity | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 11331 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 0.09% Sodium Azide | |
| PHB2 | |
| KLH conjugated synthetic peptide between 225-255 amino acids from human PHB2. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction