missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progesterone R/NR3C3 Antibody (PGR/8099R), Novus Biologicals™
Rabbit Monoclonal Antibody
£214.00 - £377.00
Specifications
| Antigen | Progesterone R/NR3C3 |
|---|---|
| Clone | PGR/8099R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18634114
|
Novus Biologicals
NBP3-23941-100ug |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18659983
|
Novus Biologicals
NBP3-23941-20ug |
20 μg |
£214.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Progesterone R/NR3C3 Monoclonal antibody specifically detects Progesterone R/NR3C3 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Progesterone R/NR3C3 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Immunology, Innate Immunity, Phospho Specific, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 5241 | |
| IgG κ | |
| Protein A or G purified |
| PGR/8099R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor | |
| Recombinant fragment (around aa 1-200) of human Progesterone R/NR3C3 (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title